Learn More
MSC Guardair Vacuum Generating Head, 10 Gallon, 1.5 in Inlet, Wet/Dry
Catalog No. 502837386
Encompass
This vacuum generating head is engineered for exceptional performance, delivering unparalleled vacuum suction while consuming up to 70% less compressed air than traditional models, leading to substantial operating cost savings. It operates at an ultra-quiet 88 dBA or less, making it 10-15 dB quieter than other vacuums.
  • Consumes up to 70% less compressed air
  • Operates quietly at 88 dBA or less
  • Customizable performance with three injector nozzle sets
  • Rugged, diecast aluminum, vacuum generating engine with a unique 2-stage, multi-venturi design
  • Suitable for both wet and dry applications with a liquid shutoff assembly
  • Backed by a Forever Warranty
Catalog No. 50-283-7386 Supplier MSC Supplier No. 64356611
This vacuum generating head is engineered for exceptional performance, delivering unparalleled vacuum suction while consuming up to 70% less compressed air than traditional models, leading to substantial operating cost savings. It operates at an ultra-quiet 88 dBA or less, making it 10-15 dB quieter than other vacuums.
  • Consumes up to 70% less compressed air
  • Operates quietly at 88 dBA or less
  • Customizable performance with three injector nozzle sets
  • Rugged, diecast aluminum, vacuum generating engine with a unique 2-stage, multi-venturi design
  • Suitable for both wet and dry applications with a liquid shutoff assembly
  • Backed by a Forever Warranty