This PowerQUAD drum-top vacuum head is air-powered and designed for mounting on any standard 55-gallon drum to provide high-performance wet/dry vacuum capability. It is engineered for industrial and commercial applications to vacuum chips, dust, debris, liquids, or sludge.
- Mounts on any standard drum for high-performance vacuum capability
- Engineered for vacuuming chips, dust, debris, liquids, or sludge
- Powered by compressed air, with no rotating parts or electric cords
- Generates superior vacuum suction and substantial operating cost savings
- Consumes up to 70% less compressed air
- Ultra-quiet operation at 88 dBA or less
- Features a rugged, diecast aluminum, multi-venturi vacuum generating engine
- Equipped with a liquid shutoff assembly for wet and dry applications
- Compatible with existing pneumatic vacuum components