Learn More
MSC Guardair Drum-Top Vacuum Head, 20/30 Gallon, 2 in Inlet, Wet/Dry
Supplier: MSC 64359128
The PowerQUAD™ Vacuum Generating Head mounts on 20 or 30-gallon drums, providing instant, high-performance vacuum capabilities for industrial and commercial applications. It efficiently vacuums chips, dust, debris, liquids, or sludge. Powered by compressed air, it eliminates rotating parts, motors, and electric cords, delivering superior vacuum suction with substantial operating cost savings.
- Up to 70% less compressed air consumption for significant operating cost savings.
- Quiet operation at 88 dBA or less, 10-15 dB quieter than traditional vacuums.
- Three color-coded, easy-to-change injector nozzle sets for customizable performance.
- Rugged, diecast aluminum engine with unique 2-stage, multi-venturi design.
- Liquid shutoff assembly for both wet and dry applications.
- Forever Warranty.