Learn More
MSC Guardair PowerQUAD Vacuum Head Kit, 20/30 Gallon, Wet/Dry
Supplier: MSC 72077043
The PowerQUAD vacuum generating head kit with a 2 in inlet offers high-performance vacuum capability when mounted on a 20 or 30-gallon drum. This pneumatic vacuum, powered by compressed air, eliminates the need for motors that can spark or burn out and has no electric cords. Engineered for industrial and commercial applications, it effectively vacuums chips, dust, debris, liquids, or sludge.
- Consumes up to 70% less compressed air, delivering unparalleled vacuum suction and significant operating cost savings.
- Operates quietly at 88 dBA or less.
- Performance can be tailored using three color-coded, easy-to-change injector nozzle sets.
- Features a rugged, diecast aluminum, vacuum generating engine with a unique 2-stage, multi-venturi design.
- Equipped with a liquid shutoff assembly, making it suitable for both wet and dry applications.