Learn More
MSC Guardair Vacuum Generating Head Kit, 10 Gallon, 1.5 in, Wet/Dry
Catalog No. 502838460
Encompass
This vacuum generating head kit is engineered for high performance in industrial and commercial applications, handling chips, dust, debris, liquids, or sludge. Powered by compressed air, it eliminates rotating parts, motors, and electric cords. It mounts on a standard 10-gallon drum for instant, high-performance vacuum capability.
  • Consumes up to 70% less compressed air, providing superior vacuum suction and operating cost savings.
  • Operates ultra-quietly at 88 dBA or less, 10-15 dB quieter than traditional vacuums.
  • Includes three color-coded injector nozzle sets to customize air savings and vacuum suction.
  • Features a rugged, diecast aluminum, 2-stage, multi-venturi vacuum generating engine.
  • Equipped with a liquid shutoff assembly for wet and dry applications.
  • Backed by an exclusive forever warranty.
  • Assembled in the USA.
Catalog No. 50-283-8460 Supplier MSC Supplier No. 64357015
This vacuum generating head kit is engineered for high performance in industrial and commercial applications, handling chips, dust, debris, liquids, or sludge. Powered by compressed air, it eliminates rotating parts, motors, and electric cords. It mounts on a standard 10-gallon drum for instant, high-performance vacuum capability.
  • Consumes up to 70% less compressed air, providing superior vacuum suction and operating cost savings.
  • Operates ultra-quietly at 88 dBA or less, 10-15 dB quieter than traditional vacuums.
  • Includes three color-coded injector nozzle sets to customize air savings and vacuum suction.
  • Features a rugged, diecast aluminum, 2-stage, multi-venturi vacuum generating engine.
  • Equipped with a liquid shutoff assembly for wet and dry applications.
  • Backed by an exclusive forever warranty.
  • Assembled in the USA.