Learn More
MSC Guardair Drum-Top Vacuum Head, Air Powered, 55 gal, 1 ea
Catalog No. 502869665
Encompass
The PowerQUAD Vacuum Generating Head mounts on standard drums for high-performance vacuum capability. Designed for industrial and commercial applications, it effectively vacuums chips, dust, debris, liquids, or sludge. Powered by compressed air, it eliminates motors, sparks, and electric cords, providing superior suction and operating cost savings.
  • Consumes up to 70% less compressed air, delivering unparalleled vacuum suction and cost savings
  • Operates quietly at 88 dBA or less
  • Features three color-coded, easy-to-change injector nozzle sets for customizable performance
  • Rugged, diecast aluminum, 2-stage, multi-venturi vacuum generating engine
  • Equipped with a liquid shutoff assembly for wet and dry applications
  • Backwards compatible with existing pneumatic vacuum components
Catalog No. 50-286-9665 Supplier MSC Supplier No. 52892502
The PowerQUAD Vacuum Generating Head mounts on standard drums for high-performance vacuum capability. Designed for industrial and commercial applications, it effectively vacuums chips, dust, debris, liquids, or sludge. Powered by compressed air, it eliminates motors, sparks, and electric cords, providing superior suction and operating cost savings.
  • Consumes up to 70% less compressed air, delivering unparalleled vacuum suction and cost savings
  • Operates quietly at 88 dBA or less
  • Features three color-coded, easy-to-change injector nozzle sets for customizable performance
  • Rugged, diecast aluminum, 2-stage, multi-venturi vacuum generating engine
  • Equipped with a liquid shutoff assembly for wet and dry applications
  • Backwards compatible with existing pneumatic vacuum components