Learn More
MSC Guardair Wet/Dry Vacuum Kit, Air-powered, 10 Gallon, 1.25 in Inlet
Catalog No. 502838458
Encompass
This pneumatic wet/dry vacuum is designed for industrial and commercial use, powered by compressed air. It eliminates electric cords and motors, generating superior vacuum suction and offering significant operating cost savings. Versatile for vacuuming chips, dust, debris, liquids, or sludge, the kit includes a vacuum hose, air supply hose, and various accessories for comprehensive cleaning tasks.
  • Consumes up to 70% less compressed air with high vacuum suction and operating cost savings.
  • Operates ultra-quietly at 88 dBA or less.
  • Customizable performance with three color-coded injector nozzle sets.
  • Rugged, diecast aluminum vacuum generating engine with a 2-stage, multi-venturi design.
  • Suitable for wet and dry applications due to its liquid shutoff assembly.
Catalog No. 50-283-8458 Supplier MSC Supplier No. 64356264
This pneumatic wet/dry vacuum is designed for industrial and commercial use, powered by compressed air. It eliminates electric cords and motors, generating superior vacuum suction and offering significant operating cost savings. Versatile for vacuuming chips, dust, debris, liquids, or sludge, the kit includes a vacuum hose, air supply hose, and various accessories for comprehensive cleaning tasks.
  • Consumes up to 70% less compressed air with high vacuum suction and operating cost savings.
  • Operates ultra-quietly at 88 dBA or less.
  • Customizable performance with three color-coded injector nozzle sets.
  • Rugged, diecast aluminum vacuum generating engine with a 2-stage, multi-venturi design.
  • Suitable for wet and dry applications due to its liquid shutoff assembly.