Learn More
MSC Guardair Wet/Dry Vacuum, Air-Powered, 55 gal, Dual Filter, 1 ea
Catalog No. 502866841
Encompass
The PowerQUAD Vacuum Family is engineered for exceptional performance in industrial and commercial applications, suitable for vacuuming chips, dust, debris, liquids, or sludge. These vacuums are powered by compressed air, eliminating the need for rotating parts, motors that can spark or burn out, and electric cords.
  • Generates superior vacuum suction and offers substantial operating cost savings
  • Operates quietly at 88 dBA or less
  • Consumes up to 70% less compressed air
  • Features a rugged, diecast aluminum, vacuum generating engine with a unique 2-stage, multi-venturi design
  • Equipped with a liquid shutoff assembly for both wet and dry applications
  • Performance can be tailored using three color-coded, easy-to-change nozzle sets
  • Backwards compatible with existing pneumatic vacuum components
  • Includes vacuum hose and air supply hose
  • Forever Warranty
Catalog No. 50-286-6841 Supplier MSC Supplier No. 64360068
The PowerQUAD Vacuum Family is engineered for exceptional performance in industrial and commercial applications, suitable for vacuuming chips, dust, debris, liquids, or sludge. These vacuums are powered by compressed air, eliminating the need for rotating parts, motors that can spark or burn out, and electric cords.
  • Generates superior vacuum suction and offers substantial operating cost savings
  • Operates quietly at 88 dBA or less
  • Consumes up to 70% less compressed air
  • Features a rugged, diecast aluminum, vacuum generating engine with a unique 2-stage, multi-venturi design
  • Equipped with a liquid shutoff assembly for both wet and dry applications
  • Performance can be tailored using three color-coded, easy-to-change nozzle sets
  • Backwards compatible with existing pneumatic vacuum components
  • Includes vacuum hose and air supply hose
  • Forever Warranty