Learn More
MSC Guardair Wet/Dry Vacuum, Air Powered, 10 gal, Steel Tank
Supplier: MSC 64356603
The PowerQUAD Wet/Dry Vacuum is an air-powered industrial vacuum with a 10-gallon steel tank, engineered for exceptional performance in industrial and commercial cleaning. It efficiently vacuums chips, dust, debris, liquids, or sludge. Its compressed air power eliminates motors and cords, providing superior suction and operating cost savings.
- Ideal for industrial and commercial cleaning of chips, dust, debris, liquids, or sludge.
- Powered by compressed air, eliminating motors and electric cords.
- Generates superior vacuum suction while consuming up to 70% less compressed air.
- Operates quietly at 88 dBA or less.
- Features a rugged, diecast aluminum engine with a 2-stage, multi-venturi design.
- Equipped with a liquid shutoff assembly for wet and dry applications.
- Backwards compatible with existing pneumatic vacuum components.