Learn More
METTLER TOLEDO 3P INDUSTRIAL POWERSHOWER FLEX
Catalog No. NC1057200
Encompass
  • Facilitates the upgrade and installation of the PowerShower™ unit.
  • Enhances rinsing capabilities for improved analytical efficiency.
  • Minimizes cross-contamination in UV/Vis spectrophotometry applications.
  • Compatible with InMotion™ Flex Autosampler system.
  • Provides proper connection for seamless operation.
  • Designed as a critical accessory for laboratory setups.
  • Supports analytical reproducibility.
Catalog No. NC1057200 Supplier METTLER TOLEDO 3P INDUSTRIAL Supplier No. 30094167
  • Facilitates the upgrade and installation of the PowerShower™ unit.
  • Enhances rinsing capabilities for improved analytical efficiency.
  • Minimizes cross-contamination in UV/Vis spectrophotometry applications.
  • Compatible with InMotion™ Flex Autosampler system.
  • Provides proper connection for seamless operation.
  • Designed as a critical accessory for laboratory setups.
  • Supports analytical reproducibility.