Learn More
MSC PowerSmith Rechargeable Foldable LED Inspection Light, 600 Lumens, 1 ea
Catalog No. 5000782945
Encompass
This rechargeable and foldable LED inspection light features multiple functions, including a spot light, flood light, and UV light. It has a foldable and rotatable light panel for greater versatility.
  • Multiple functions: spot light, flood light, and UV light
  • Foldable and rotatable light panel
  • Dimmable 200 lumens spot light for distance up to 164 ft.
  • Dimmable 600 lumens flood light for lighting larger area
  • UV light with 395nm wavelength for detection of fluorescent agents, security codes, and stains
  • Sealed on/off & mode switch with battery power indicator
  • Powered by rechargeable 3.7V, 2.0Ah lithium-ion battery
  • USB power output for charging other portable devices
  • Magnetic base, metal hanging hook, and spring-loaded mounting clamp
  • IP43 for indoor/outdoor use
  • Includes USB-C charging cable
Catalog No. 50-007-82945 Supplier MSC Supplier No. 95507208
This rechargeable and foldable LED inspection light features multiple functions, including a spot light, flood light, and UV light. It has a foldable and rotatable light panel for greater versatility.
  • Multiple functions: spot light, flood light, and UV light
  • Foldable and rotatable light panel
  • Dimmable 200 lumens spot light for distance up to 164 ft.
  • Dimmable 600 lumens flood light for lighting larger area
  • UV light with 395nm wavelength for detection of fluorescent agents, security codes, and stains
  • Sealed on/off & mode switch with battery power indicator
  • Powered by rechargeable 3.7V, 2.0Ah lithium-ion battery
  • USB power output for charging other portable devices
  • Magnetic base, metal hanging hook, and spring-loaded mounting clamp
  • IP43 for indoor/outdoor use
  • Includes USB-C charging cable