Learn More
ILEX LIFE SCIENCES LLC POWERSTEM MSC1 SFM KIT 500 ML
SDP
Supplier:  ILEX LIFE SCIENCES LLC P0477355K
Encompass_Preferred
PAN-Biotech PowerStem MSC1, Serum-free medium kit for human mesenchymal stem cells (hMSC), 500 ml kit. MANUFACTURER: PAN-Biotech GmbH. COUNTRY OF ORIGIN: Germany. INTRO: PAN-Biotech PowerStem MSC1 is an easy to use xeno-free, serum-free medium for in vitro proliferation of human mesenchymal stem cells (hMSC). PowerStem MSC1 supports long-term growth of MSC and preserves their multi-lineage potential. In addition, MSC cultured in PowerStem MSC1 expands faster and shows a significant reduction in hematopoietic cell contamination at early passages compared to serum-based media. To differentiate the proliferated MSC into different cells types the relevant protocols and differentiation factors should be used. COMPOSITION: PowerStem MSC1 contains salts, amino acids, trace elements, hormones, growth factors, enriched human proteins and lipids in an optimized formulation. COMPONENTS: 470 ml Basal Medium, 5 ml Growth Supplement 1, 25 ml Growth Supplement 2.
Catalog No. 50-273-3770