Learn More
Glas Col LLC Control
SDP
Catalog No. NC0194512
Encompass_Preferred
PowrTrol proportional voltage control, from 5-100% of rated voltage, 10A, 1200W, 120V
Catalog No. NC0194512 Supplier Glas Col LLC Supplier No. 104A PL120
PowrTrol proportional voltage control, from 5-100% of rated voltage, 10A, 1200W, 120V