Learn More
Glas Col LLC Control
SDP
Catalog No. NC1134702
Encompass_Preferred
PowrTwin proportional voltage controller. Capable of controlling two separate circuits. 5-100% adjustable, 7A, 2 circuits @ 840W, 120V. Precise manual control.
Catalog No. NC1134702 Supplier Glas Col LLC Supplier No. 104A PL1202
PowrTwin proportional voltage controller. Capable of controlling two separate circuits. 5-100% adjustable, 7A, 2 circuits @ 840W, 120V. Precise manual control.