Learn More
Sigma Aldrich Fine Chemicals Biosciences Poxygrid Centrifuge Bottle Rack, each
Catalog No. NC1255718
Encompass
The only rack that can support up to four (4) 500ml conical bottom centrifuge tubes that cannot stand upright by themselves.
Catalog No. NC1255718 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BAF1985605001EA
The only rack that can support up to four (4) 500ml conical bottom centrifuge tubes that cannot stand upright by themselves.