Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bel-Art™ Bench-Top Biohazard Bag Holder
GSA_VASCI_ED

Catalog No. S50397A Shop All SP Bel-Art & SP Wilmad-LabGlass Products
Change view
Click to view available options
Product Type:
Bag Holder Cover
Biohazard Bag Holder
Biohazard Bag Holder Kit
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S50397A Biohazard Bag Holder
S04788 Bag Holder Cover
S04788B Biohazard Bag Holder Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. S50397A Supplier Bel-Art™ Supplier No. H131931000/EMD
Get Up to $500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Includes

Rectangular stainless steel wire frame only

Economical and convenient, this benchtop disposal system is perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use

  • Rectangular steel wire frame to hold one polyethylene bag of dimensions 8.5 x 11 in. (22 x 28 cm)
  • Available with epoxy coat in orange color
  • Comes with snug fitting polypropylene plastic cover to hold bag securely in place
  • Reusable and allows cleaning by hand or in automated dish washers
  • Autoclavable
  • Also used with Clavies Autoclavable Bags H13182-0812

Order Info

For the bag holder, cover and biohazard bags, use Cat. No. S04788B.

Compliance

When used with lid, provides compliance with 29 CFR 1910.1030(d)(2)(xiii) (A), which requires a cover on any container used to store, transport or ship biohazardous wastes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Autoclavable Autoclavable
Color Orange
Disposable Single-use
For Use With (Application) Perfect for collecting pipette tips, microcentrifuge tubes, etc. during daily use
For Use With (Equipment) 01-828A, 01-828-031, 01-830A, 01-826-1, 01-827A, 01-829A, 01-815 11 H x 8.5 in. W Biohazard Bags biohazard bag, Clavies autoclavable bags (01-815-6)
Includes Rectangular stainless steel wire frame only
Material Epoxy-coated Steel Wire
Dimensions (L x W x H) 3.4 x 5 x 8.5 in. (8.63 x 12.7 x 21.59 cm)
Length (English) 3.4 in.
Width (English) 5 in.
Height (English) 8.5 in.
Height (Metric) 21.59 cm
Product Type Biohazard Bag Holder
Length (Metric) 8.63 cm
Width (Metric) 12.7 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.