Learn More
Alkali Scientific Poxygrid "Half-Size" Test Tube Rack, 16-20mm, Blue, 20 Places
SDP
Catalog No. 502388806
Encompass_Preferred
Durable and smooth coating is impervious to organic solvents, salts, weak acids and weak alkalis. Poxygrid "half-size" is an ideal space saver, perfect for use in water baths, under fume hoods, and other limited space locations. Holds 16-20mm tubes in a 5 x 4 configuration. Epoxy coated steel wire racks are chemical and corrosion resistant. Withstands temperatures ranging from -223C (-369F) to 121C (250F) and can be autoclaved. One per pack.
Catalog No. 50-238-8806 Supplier Alkali Scientific Supplier No. R8120
Durable and smooth coating is impervious to organic solvents, salts, weak acids and weak alkalis. Poxygrid "half-size" is an ideal space saver, perfect for use in water baths, under fume hoods, and other limited space locations. Holds 16-20mm tubes in a 5 x 4 configuration. Epoxy coated steel wire racks are chemical and corrosion resistant. Withstands temperatures ranging from -223C (-369F) to 121C (250F) and can be autoclaved. One per pack.