Learn More
Sigma Aldrich Fine Chemicals Biosciences PoxygridR Centrifuge Bottl1EA
Catalog No. 502901067
Encompass
Each well is 60mm square to hold bottles. Epoxy coated steel wire is chemical and corrosion resistant and steam autoclavable at 121AºC (250Aºf). Organize transport and store conical and round bottom centrifuge bottles.
Catalog No. 50-290-1067 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. BAF1985602501EA
Each well is 60mm square to hold bottles. Epoxy coated steel wire is chemical and corrosion resistant and steam autoclavable at 121AºC (250Aºf). Organize transport and store conical and round bottom centrifuge bottles.