Learn More
Selleck Chemical LLC Poziotinib
SDP
Catalog No. 5000004588
Encompass_Preferred
Poziotinib is an irreversible pan-HER inhibitor with IC50 of 3 2 nM 5 3 nM and 23 5 nM for HER1 HER2 and HER4 respectively Poziotinib also induces apoptosis and G1 cell cycle arrest Phase 2
Catalog No. 50-000-04588 Supplier Selleck Chemical LLC Supplier No. S7358100MG
Poziotinib is an irreversible pan-HER inhibitor with IC50 of 3 2 nM 5 3 nM and 23 5 nM for HER1 HER2 and HER4 respectively Poziotinib also induces apoptosis and G1 cell cycle arrest Phase 2