Learn More
Selleck Chemical LLC Poziotinib (HM781-36B) 5mg 1092364-38-9 NOV120101
SDP
Catalog No. 501364682
Encompass_Preferred
Poziotinib (HM781-36B, NOV120101) is an irreversible pan-HER inhibitor with IC50 of 3.2 nM, 5.3 nM and 23.5 nM for HER1, HER2, and HER4, respectively. Poziotinib also induces apoptosis and G1 cell cycle arrest. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4682 Supplier Selleck Chemical LLC Supplier No. S73585MG
Poziotinib (HM781-36B, NOV120101) is an irreversible pan-HER inhibitor with IC50 of 3.2 nM, 5.3 nM and 23.5 nM for HER1, HER2, and HER4, respectively. Poziotinib also induces apoptosis and G1 cell cycle arrest. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.