Learn More
Cole-Parmer Polypropylene storage boxes with unattached lid for 50 (0.5 mL) tubes, 6/pack
Catalog No. NC9073940 Shop All Cole-Parmer Products
Encompass
Polypropylene (PP) storage boxes are sturdy and wont deteriorate when exposed to moisture and temperatures down to 80C (112F). These boxes are excellent substitutes for cardboard boxes that tend to degrade and absorb moisture after prolonged use. Translucent plastic lids let you easily identify samples. Boxes accommodate microcentrifuge tubes and cryogenic tubes of various lengths. Autoclavable.
Catalog No. NC9073940 Supplier Cole-Parmer Supplier No. 4435000
Polypropylene (PP) storage boxes are sturdy and wont deteriorate when exposed to moisture and temperatures down to 80C (112F). These boxes are excellent substitutes for cardboard boxes that tend to degrade and absorb moisture after prolonged use. Translucent plastic lids let you easily identify samples. Boxes accommodate microcentrifuge tubes and cryogenic tubes of various lengths. Autoclavable.