Learn More
Beckman Coulter Beckman Echo Qualified 384-Well Polypropylene Microplate, Clear, Non-sterile
Catalog No. NC2684864
Encompass_Preferred
The Echo-qualified 384-well polypropylene (PP) source microplate is engineered for acoustic liquid handling and compound or sample storage. Its polypropylene construction provides chemical resistance to solvents such as DMSO and common buffers while supporting precise transfer performance for screening and compound management workflows.
  • 384-well format for high-density screening and storage.
  • Polypropylene construction resists solvents including DMSO and common buffers.
  • Source microplate design suitable for acoustic liquid handling workflows.
  • Clear plate finish allows visual inspection of wells.
  • Non-sterile packaging for general laboratory storage and compound management.
Catalog No. NC2684864 Supplier Beckman Coulter Supplier No. 00114555/INTEGRATED
Add to Cart
Add to Cart
The Echo-qualified 384-well polypropylene (PP) source microplate is engineered for acoustic liquid handling and compound or sample storage. Its polypropylene construction provides chemical resistance to solvents such as DMSO and common buffers while supporting precise transfer performance for screening and compound management workflows.
  • 384-well format for high-density screening and storage.
  • Polypropylene construction resists solvents including DMSO and common buffers.
  • Source microplate design suitable for acoustic liquid handling workflows.
  • Clear plate finish allows visual inspection of wells.
  • Non-sterile packaging for general laboratory storage and compound management.