Learn More
Accuflow Systems 1"/1.5" TC x 1/4" ID Polypropylene Bio Barb Adapter
Catalog No. 501108270
Encompass_Preferred
1"/1.5" TC x 1/4" ID Polypropylene Bio Barb Adapter with true-bore ID are designed for use in GMP Manufacturing and laboratories. These Bio Barb adapters can be autoclavable as well as gama irradiated. Constructed of USP Class VI Polypropylene. Designed for Sanitary Tri clamp applications. Max working pressure, 150psi @ 70? F, Pack of 10
Catalog No. 50-110-8270 Supplier Accuflow Systems Supplier No. PP100/150TC0250BB
1"/1.5" TC x 1/4" ID Polypropylene Bio Barb Adapter with true-bore ID are designed for use in GMP Manufacturing and laboratories. These Bio Barb adapters can be autoclavable as well as gama irradiated. Constructed of USP Class VI Polypropylene. Designed for Sanitary Tri clamp applications. Max working pressure, 150psi @ 70? F, Pack of 10