Learn More
Accuflow Systems 1"/1.5" TC x 3/8" ID Polypropylene Bio Barb Adapte
Supplier:  Accuflow Systems PP100/150TC0375BB
Encompass_Preferred
1"/1.5" TC x 3/8" ID Polypropylene Bio Barb Adapter with true-bore ID are designed for use in GMP Manufacturing and laboratories. These Bio Barb adapters can be autoclavable as well as gama irradiated. Constructed of USP Class VI Polypropylene. Designed for Sanitary Tri clamp applications. Max working pressure, 150psi @ 70? F, Pack of 10
Catalog No. 50-110-8271