Learn More
Flacktek Inc SC 12 Cup (100 pcs) - Translucent
SDP
Catalog No. NC1899534
Encompass
Total Container Volume: 12 cc
Recommended Mixing Volume: (1/3-1/2 of the container volume)
Cup Material: Polypropylene (PP)
Machine Compatibility: FlackTek Series 330 up to 1200
Catalog No. NC1899534 Supplier Flacktek Inc Supplier No. 150 244
Total Container Volume: 12 cc
Recommended Mixing Volume: (1/3-1/2 of the container volume)
Cup Material: Polypropylene (PP)
Machine Compatibility: FlackTek Series 330 up to 1200