Learn More
Selleck Chemical LLC PP121 10mg 1092788-83-4
SDP
Supplier:  Selleck Chemical LLC S262210MG
Encompass_Preferred
PP-121 is a multi-targeted inhibitor of PDGFR, Hck, mTOR, VEGFR2, Src and Abl with IC50 of 2 nM, 8 nM, 10 nM, 12 nM, 14 nM and 18 nM, also inhibits DNA-PK with IC50 of 60 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-502-6