Learn More
Hach Company PP Beaker Low Form 1000mL 3/pk
Catalog No. 5000257145 Shop All Hach Company Products
Encompass
Griffin style (low form) beakers from Nalgene are the industry standard fora wide variety of lab applications Graduations are high quality screen printing in sizes 250mL Bull nose lip and molded pour spout minimize spillage in liquid transfer Autoclavable non-toxic and biologically inert Volumes and graduations are approximate
Catalog No. 50-002-57145 Supplier Hach Company Supplier No. 108083
Griffin style (low form) beakers from Nalgene are the industry standard fora wide variety of lab applications Graduations are high quality screen printing in sizes 250mL Bull nose lip and molded pour spout minimize spillage in liquid transfer Autoclavable non-toxic and biologically inert Volumes and graduations are approximate