Learn More
Bellco Glass, Inc. PP Blk Open Hole S/Cap,10mm 10-425mm
SDP
Catalog No. 501945387
Encompass_Preferred
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 10mm
Catalog No. 50-194-5387 Supplier Bellco Glass, Inc. Supplier No. 207000975
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 10mm