Learn More
Bellco Glass, Inc. PP Blk S/Cap W/PTFE Liner,13mm 13-425
SDP
Catalog No. 501945388
Encompass_Preferred
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 13mm
Catalog No. 50-194-5388 Supplier Bellco Glass, Inc. Supplier No. 207000980
Designed for standard screw thread vials. Available with or without septa. Easy on and easy off convenience with a twist of the cap that produces an evaporation proof seal. Larger opening provides a increased target area for needle penetration. MADE IN THE USA. Size: 13mm