Learn More
Bellco Glass, Inc. PP Blue Screw Cap, 32mm Solid Cap
SDP
Catalog No. 501945754
Encompass_Preferred
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 32mm Color: blue
Catalog No. 50-194-5754 Supplier Bellco Glass, Inc. Supplier No. 563700032
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 32mm Color: blue