Learn More
Bellco Glass, Inc. PP Blue Screw Cap, 45mm Solid
SDP
Catalog No. 501945759
Encompass_Preferred
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 45mm Color: blue
Catalog No. 50-194-5759 Supplier Bellco Glass, Inc. Supplier No. 563700045
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 45mm Color: blue