Learn More
AVANTOR MASTERFLEX (VWR LLC) High-Head Enclosed Motor PP Centrifugal Pump; 2.9 GPM/22.6 ft, 115V
Catalog No. NC1309833
Encompass
High-head polypropylene pumps have a closed impeller design to significantly increase the output pressure. Polypropylene construction provides excellent chemical resistance in applications, such as filtration, where a high-head is needed.
Catalog No. NC1309833 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX7201200
High-head polypropylene pumps have a closed impeller design to significantly increase the output pressure. Polypropylene construction provides excellent chemical resistance in applications, such as filtration, where a high-head is needed.