Learn More
Bellco Glass, Inc. Polypropylene Erlenmeyer Grad Flask,1000mL
SDP
Catalog No. 501945435
Encompass_Preferred
Flask have wide-mouth design for easy pouring and printed scale of approximate volumes Material: polypropylene Includes: polypropylene screw cap Size: 1000ml Volume: 1000 x 200mm Thread: 52mm
Catalog No. 50-194-5435 Supplier Bellco Glass, Inc. Supplier No. 250001000
Flask have wide-mouth design for easy pouring and printed scale of approximate volumes Material: polypropylene Includes: polypropylene screw cap Size: 1000ml Volume: 1000 x 200mm Thread: 52mm