Learn More
Bellco Glass, Inc. Polypropylene Erlenmeyer Grad Flask,100mL
SDP
Catalog No. 501945432
Encompass_Preferred
Flask have wide-mouth design for easy pouring and printed scale of approximate volumes Material: polypropylene Includes: polypropylene screw cap Size: 100ml Volume: 100 x 20mm Thread: 40mm
Catalog No. 50-194-5432 Supplier Bellco Glass, Inc. Supplier No. 250000100
Flask have wide-mouth design for easy pouring and printed scale of approximate volumes Material: polypropylene Includes: polypropylene screw cap Size: 100ml Volume: 100 x 20mm Thread: 40mm