Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC λPP protein, Escherichia phage lambda (His, FLAG) | >85.0% | 10UG
SDP

Catalog No. 5000176079
Encompass_Preferred

Recombinant λPP protein from Escherichia phage lambda, expressed in Escherichia coli with N-terminal 6xHis and FLAG affinity tags. Supplied as a 0.22 μm filtered solution formulated for stability and downstream biochemical assays.

  • Recombinant protein expressed in Escherichia coli for consistent lot-to-lot quality.
  • N-terminal 6xHis and FLAG tags for affinity purification and detection.
  • Partial-length protein with molecular weight 27.2 kDa.
  • Purity >85.0% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in 50 mM Tris-HCl pH 7.5, 200 mM NaCl, 20% glycerol (0.22 μm filtered).
  • Supplied as a 10 μg unit, shipped on dry ice and stored at -80°C.
  • Rapid thawing recommended; avoid repeated freeze-thaw cycles.

Catalog No. 50-001-76079 Supplier Medchemexpress LLC Supplier No. HYP70199210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant λPP protein from Escherichia phage lambda, expressed in Escherichia coli with N-terminal 6xHis and FLAG affinity tags. Supplied as a 0.22 μm filtered solution formulated for stability and downstream biochemical assays.

  • Recombinant protein expressed in Escherichia coli for consistent lot-to-lot quality.
  • N-terminal 6xHis and FLAG tags for affinity purification and detection.
  • Partial-length protein with molecular weight 27.2 kDa.
  • Purity >85.0% as determined by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Formulated in 50 mM Tris-HCl pH 7.5, 200 mM NaCl, 20% glycerol (0.22 μm filtered).
  • Supplied as a 10 μg unit, shipped on dry ice and stored at -80°C.
  • Rapid thawing recommended; avoid repeated freeze-thaw cycles.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.