Learn More
Medchemexpress LLC LambdaPP protein, Escherichia phage lambda (His, FLAG) | 95.0% | 50UG
SDP
Catalog No. 5000174382
Encompass_Preferred
Recombinant lambdaPP protein from Escherichia phage lambda, expressed in E. coli and supplied with N-terminal 6xHis and N-terminal FLAG affinity tags. The protein is approximately 220 amino acids in length, purified to ~95.0%, and intended for use in biochemical applications such as electrophoresis and western blotting. Supplied in small research quantities and stored at -80 °C for stability.
  • Contains N-terminal 6xHis and N-terminal FLAG affinity tags for straightforward purification and detection.
  • Expressed in Escherichia coli to provide high-yield recombinant protein.
  • Purified to 95.0% to minimize contaminants in downstream assays.
  • Approximate length of 220 amino acids for predictable behavior in assays.
  • Suitable for electrophoresis and western blot applications.
  • Stored at -80 °C to maintain stability for up to one year.
Catalog No. 50-001-74382 Supplier Medchemexpress LLC Supplier No. HYP70199250UG
Recombinant lambdaPP protein from Escherichia phage lambda, expressed in E. coli and supplied with N-terminal 6xHis and N-terminal FLAG affinity tags. The protein is approximately 220 amino acids in length, purified to ~95.0%, and intended for use in biochemical applications such as electrophoresis and western blotting. Supplied in small research quantities and stored at -80 °C for stability.
  • Contains N-terminal 6xHis and N-terminal FLAG affinity tags for straightforward purification and detection.
  • Expressed in Escherichia coli to provide high-yield recombinant protein.
  • Purified to 95.0% to minimize contaminants in downstream assays.
  • Approximate length of 220 amino acids for predictable behavior in assays.
  • Suitable for electrophoresis and western blot applications.
  • Stored at -80 °C to maintain stability for up to one year.