Learn More
Analytical Sales & Services PP Ferrule for 1/16" OD Tubing
Catalog No. 5000805065
Encompass
Natural Polypropylene Flangeless Ferrule for 1/16" OD Tubing
Qty/Pkg: EACH
Catalog No. 50-008-05065 Supplier Analytical Sales & Services Supplier No. 59221
Natural Polypropylene Flangeless Ferrule for 1/16" OD Tubing
Qty/Pkg: EACH