Learn More
Analytical Sales & Services PP Flangeless Nut for 1/16" OD Tubing
Catalog No. 5000804669
Encompass
1/4-28 Flangeless Polypropylene Male Nut for 1/16" OD Tubing
Qty/Pkg: 10PK
Catalog No. 50-008-04669 Supplier Analytical Sales & Services Supplier No. 59220X
1/4-28 Flangeless Polypropylene Male Nut for 1/16" OD Tubing
Qty/Pkg: 10PK