Learn More
Bellco Glass, Inc. PP Green Screw Cap,45mm Solid Cap
SDP
Catalog No. 501945761
Encompass_Preferred
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 45mm Color: green
Catalog No. 50-194-5761 Supplier Bellco Glass, Inc. Supplier No. 563700047
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 45mm Color: green