Learn More
Bellco Glass, Inc. PP Grey Screw Cap,45mm Solid
SDP
Catalog No. 501945763
Encompass_Preferred
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 45mm Color: Grey
Catalog No. 50-194-5763 Supplier Bellco Glass, Inc. Supplier No. 563700049
Molded from Polypropylene with inner ring that seals into the neck of the bottle. They are autoclavable to 140 degrees Celsius. MADE IN THE USA Size: 45mm Color: Grey