Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,1000mL
SDP
Catalog No. 501945723
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 1000mL x 100mL Dimensions:102x150mm
Catalog No. 50-194-5723 Supplier Bellco Glass, Inc. Supplier No. 562301000
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 1000mL x 100mL Dimensions:102x150mm