Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,100mL
SDP
Catalog No. 501945716
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 100ml x 20mL Dimensions: 50x70mm
Catalog No. 50-194-5716 Supplier Bellco Glass, Inc. Supplier No. 562300100
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 100ml x 20mL Dimensions: 50x70mm