Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,10mL
SDP
Catalog No. 501945713
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 10ml x 2mL Dimensions: 25x35mm
Catalog No. 50-194-5713 Supplier Bellco Glass, Inc. Supplier No. 562300010
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 10ml x 2mL Dimensions: 25x35mm