Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,2000mL
SDP
Catalog No. 501945724
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 2000mL x 200mL Dimensions: 133x183mm
Catalog No. 50-194-5724 Supplier Bellco Glass, Inc. Supplier No. 562302000
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 2000mL x 200mL Dimensions: 133x183mm