Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,250mL
SDP
Catalog No. 501945718
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 250ml x 50mL Dimensions: 68x94mm
Catalog No. 50-194-5718 Supplier Bellco Glass, Inc. Supplier No. 562300250
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 250ml x 50mL Dimensions: 68x94mm