Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,25mL
SDP
Catalog No. 501945714
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 25ml x 5mL Dimensions: 31x47mm
Catalog No. 50-194-5714 Supplier Bellco Glass, Inc. Supplier No. 562300025
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 25ml x 5mL Dimensions: 31x47mm