Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,3000mL
SDP
Catalog No. 501945725
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 3000mL x 500mL Dimensions: 174x214mm
Catalog No. 50-194-5725 Supplier Bellco Glass, Inc. Supplier No. 562303000
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 3000mL x 500mL Dimensions: 174x214mm