Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,400mL
SDP
Catalog No. 501945719
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 400ml x 50mL Dimensions: 77x109mm
Catalog No. 50-194-5719 Supplier Bellco Glass, Inc. Supplier No. 562300400
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 400ml x 50mL Dimensions: 77x109mm