Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,5000mL
SDP
Catalog No. 501945726
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 5000mL x 500mL Dimensions: 185x248mm
Catalog No. 50-194-5726 Supplier Bellco Glass, Inc. Supplier No. 562305000
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 5000mL x 500mL Dimensions: 185x248mm