Learn More
Bellco Glass, Inc. Polypropylene Griffin Grad Beaker,500mL
SDP
Catalog No. 501945720
Encompass
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 500ml x 50mL Dimensions: 88x122mm
Catalog No. 50-194-5720 Supplier Bellco Glass, Inc. Supplier No. 562300500
Add to Cart
Add to Cart
These beakers are useful for general laboratory procedures and are autoclaveable. They have blue imprinted graduations to aid in viewing and dispensing liquids. Material: polypropylene Size: 500ml x 50mL Dimensions: 88x122mm